Learn More
MSC Dow Corning 99180646 10.1 oz Cartridge White RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Catalog No. 501305369
Encompass
Dow Corning 99180646 10.1 oz Cartridge White RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737
Catalog No. 50-130-5369 Supplier MSC Supplier No. 00231944
Dow Corning 99180646 10.1 oz Cartridge White RTV Silicone Joint Sealant -85 to 350°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 737