Learn More
Ecolab Ecolab Oasis Neutral Disinfectant Cleaner, Germicidal, Deodorizes, 2.5 Gallons
Catalog No. NC3302440
Encompass
Ecolab Oasis Neutral Disinfectant Cleaner is a concentrated, multi-purpose solution designed for effective disinfection, cleaning, and deodorizing of hard, non-porous surfaces. It excels in hard water conditions up to 200 ppm and provides a clean, fresh scent. With germicidal, fungicidal, and mildewstatic properties, it ensures comprehensive microbial control in various environments, making maintenance efficient and effective.
Catalog No. NC3302440 Supplier Ecolab Supplier No. 6101205
Ecolab Oasis Neutral Disinfectant Cleaner is a concentrated, multi-purpose solution designed for effective disinfection, cleaning, and deodorizing of hard, non-porous surfaces. It excels in hard water conditions up to 200 ppm and provides a clean, fresh scent. With germicidal, fungicidal, and mildewstatic properties, it ensures comprehensive microbial control in various environments, making maintenance efficient and effective.