Learn More
Bioworld Neutral Dissociator, pH 7.0, 50 mL
Catalog No. 50198512
Encompass_Preferred
Neutral Dissociator, pH 7.0 Two reagents are provided to dissociate your antigen-antibody complex in a column. Dialyze to desalt after dissociating under denaturing conditions with neutral dissociator.
Catalog No. 50-198-512 Supplier Bioworld Supplier No. 208800001
Neutral Dissociator, pH 7.0 Two reagents are provided to dissociate your antigen-antibody complex in a column. Dialyze to desalt after dissociating under denaturing conditions with neutral dissociator.