Learn More
Syngene™ Neutral Field Screen Set for use with G:BOX Systems
GREENER_CHOICESDP
Catalog No. 01257272 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
G:BOX systems
Catalog No. For Use With (Equipment)
01-257-272 G:BOX systems
1 options
Catalog No. 01-257-272 Supplier Syngene™ Supplier No. GXNFS
Add to Cart
Add to Cart
Syngene™ Neutral Field Screen Set for use with G:BOX Systems that includes a blue screen and a frosted screen.
  • Corrects for any uneven Illumination by acquiring a neutral-field reference image with the same ntensity illumination as the experiment
  • For use with Epi-UV lighting and transilluminators
Specifications
For Use With (Equipment) G:BOX systems
Includes 1 blue screen and 1 frosted screen
Dimensions (L x W x H) 335 x 330 x 15 mm