Learn More
MSC Detco Floor Cleaner, Concentrated, Neutral, 55 Gal Drum
Catalog No. 502839032
Encompass
This Kwik Mop is a concentrated neutral floor cleaner supplied in a 55-gallon drum. It is biodegradable and designed for use on resilient flooring, maintaining a pH of 7.00.
  • Concentrated formula for effective cleaning
  • Neutral pH of 7.00, suitable for various floor types
  • Ideal for resilient flooring applications
  • Supplied in a 55-gallon drum for bulk use
  • Biodegradable for environmental consideration
Catalog No. 50-283-9032 Supplier MSC Supplier No. 53244489
This Kwik Mop is a concentrated neutral floor cleaner supplied in a 55-gallon drum. It is biodegradable and designed for use on resilient flooring, maintaining a pH of 7.00.
  • Concentrated formula for effective cleaning
  • Neutral pH of 7.00, suitable for various floor types
  • Ideal for resilient flooring applications
  • Supplied in a 55-gallon drum for bulk use
  • Biodegradable for environmental consideration