Learn More
Enzo Life Sciences Neutral-ODN (Control for iODNs) (endotoxin-free) (synthetic) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX746256C100
Encompass_Preferred
Inactive (neutral) control ODN for in vivo use in rodents (50-150µg per injection) without agonistic nor antagonistic activity. Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9678