Learn More
Medchemexpress LLC Neutral protease, Paenibacillus polymyxa | 42613-33-2 | 33.2 kDa (monomer); 66.3 kDa (dimer) | Not applicable - protein (no single chemical formula) | 5 MG
SDP
Catalog No. 5000262811
Encompass_Preferred
Neutral protease, Paenibacillus polymyxa is a bacterial neutral metalloprotease used in research to selectively cleave extracellular matrix proteins such as fibronectin and type IV collagen. Supplied as a white to off-white solid, it is commonly used for tissue dissociation, separation of epidermis from dermis, and release of intact epithelial sheets in culture. Refer to the certificate of analysis for recommended storage and specific activity data.
  • Neutral metalloprotease active against fibronectin and type IV collagen.
  • Suitable for tissue dissociation and separation of epidermis from dermis.
  • Enzyme activity ≥ 0.5 U/mg (solid), unit definition provided.
  • White to off-white solid formulation for laboratory use.
  • Shipped at room temperature; follow COA for storage conditions.
Catalog No. 50-002-62811 Supplier Medchemexpress LLC Supplier No. HY1315775MG
Neutral protease, Paenibacillus polymyxa is a bacterial neutral metalloprotease used in research to selectively cleave extracellular matrix proteins such as fibronectin and type IV collagen. Supplied as a white to off-white solid, it is commonly used for tissue dissociation, separation of epidermis from dermis, and release of intact epithelial sheets in culture. Refer to the certificate of analysis for recommended storage and specific activity data.
  • Neutral metalloprotease active against fibronectin and type IV collagen.
  • Suitable for tissue dissociation and separation of epidermis from dermis.
  • Enzyme activity ≥ 0.5 U/mg (solid), unit definition provided.
  • White to off-white solid formulation for laboratory use.
  • Shipped at room temperature; follow COA for storage conditions.