Learn More
Medchemexpress LLC Neutral Red | 553-24-2 | 98.1% | 1 ML
SDP
Catalog No. 5000410022
Encompass_Preferred
Neutral Red is a nitrogenous pH-indicator with a pKi of 6.8, acting as an indicator for the internal acidification of thylakoids and staining lysosomes red. It is used in cell cultures for plate titration of viruses.
  • As cells begin to die, their ability to incorporate neutral red diminishes
  • Fluorescent dye
  • Used for biology analysis
  • Used for microbiological analysis
  • Used for plant analysis
  • Used for bacteriological dyes
Catalog No. 50-004-10022 Supplier Medchemexpress LLC Supplier No. HYD016610MM/1ML
Neutral Red is a nitrogenous pH-indicator with a pKi of 6.8, acting as an indicator for the internal acidification of thylakoids and staining lysosomes red. It is used in cell cultures for plate titration of viruses.
  • As cells begin to die, their ability to incorporate neutral red diminishes
  • Fluorescent dye
  • Used for biology analysis
  • Used for microbiological analysis
  • Used for plant analysis
  • Used for bacteriological dyes