Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutral Red, Certified, Sigma-Aldrich

Catalog No. 01259073 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-073 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-073 Supplier MilliporeSigma Supelco Supplier No. 86125125G
Add to Cart
Add to Cart

Useful as an indicator for preparing neutral red paper, and as a biological stain.

TRUSTED_SUSTAINABILITY

Specifications

CAS 553-24-2
Molecular Weight (g/mol) 288.78 g/mol
Grade Certified
Color Dark Brown
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.