Learn More
Alkali Scientific Neutral Red Dye, 100 G
SDP
Catalog No. 5000661475
Encompass_Preferred
Neutral Red is a vital dye used for cell viability staining in biological and medical research. This 100g bottle offers an ample quantity of dye that can be used for a variety of applications, including cell culture studies, cytotoxicity assays, and tissue section staining. Neutral Red is particularly useful for assessing the viability of cultured cells, as it is absorbed by viable cells but excluded from dead cells. This ability to distinguish between living and dead cells makes Neutral Red a valuable tool in research focused on cellular health, drug testing, and toxicity studies. It is widely used in cell biology, microbiology, and pharmaceutical research.
Catalog No. 50-006-61475 Supplier Alkali Scientific Supplier No. NEU001CP
Neutral Red is a vital dye used for cell viability staining in biological and medical research. This 100g bottle offers an ample quantity of dye that can be used for a variety of applications, including cell culture studies, cytotoxicity assays, and tissue section staining. Neutral Red is particularly useful for assessing the viability of cultured cells, as it is absorbed by viable cells but excluded from dead cells. This ability to distinguish between living and dead cells makes Neutral Red a valuable tool in research focused on cellular health, drug testing, and toxicity studies. It is widely used in cell biology, microbiology, and pharmaceutical research.