Learn More
Alkali Scientific Neutral Red Dye, 25 G
SDP
Catalog No. 5000661329
Encompass_Preferred
Neutral Red Dye is used as a cell viability stain, allowing researchers to determine the health of cells in culture. This 25g bottle is ideal for laboratories conducting routine staining and viability assessments in cellular biology studies. Neutral Red is absorbed by living cells but not by dead ones, making it a useful marker for distinguishing between viable and non-viable cells. It is commonly used in cytotoxicity assays, tissue culture, and microbiology studies to assess the effects of different treatments on cell health. The dye is widely used for evaluating cellular health and is a standard reagent in laboratory procedures requiring live/dead cell differentiation.
Catalog No. 50-006-61329 Supplier Alkali Scientific Supplier No. NEU002CP
Neutral Red Dye is used as a cell viability stain, allowing researchers to determine the health of cells in culture. This 25g bottle is ideal for laboratories conducting routine staining and viability assessments in cellular biology studies. Neutral Red is absorbed by living cells but not by dead ones, making it a useful marker for distinguishing between viable and non-viable cells. It is commonly used in cytotoxicity assays, tissue culture, and microbiology studies to assess the effects of different treatments on cell health. The dye is widely used for evaluating cellular health and is a standard reagent in laboratory procedures requiring live/dead cell differentiation.