Learn More
Alkali Scientific Neutral Red Dye, 5 G
SDP
Catalog No. 5000660892
Encompass_Preferred
Neutral Red Dye, available in a 5g container, is widely used for staining living cells in biological and medical research. This dye selectively accumulates in viable cells, allowing researchers to distinguish between living and dead cells in cell culture and cytotoxicity assays. Neutral Red is ideal for applications requiring cell viability testing, such as drug testing, toxicology studies, and cell-based assays. It is frequently used in microbiology, cytology, and pharmaceutical research to monitor the effects of various substances on cell health. The 5g bottle is a convenient size for smaller scale studies or for labs with less frequent staining requirements.
Catalog No. 50-006-60892 Supplier Alkali Scientific Supplier No. NEU003CP
Neutral Red Dye, available in a 5g container, is widely used for staining living cells in biological and medical research. This dye selectively accumulates in viable cells, allowing researchers to distinguish between living and dead cells in cell culture and cytotoxicity assays. Neutral Red is ideal for applications requiring cell viability testing, such as drug testing, toxicology studies, and cell-based assays. It is frequently used in microbiology, cytology, and pharmaceutical research to monitor the effects of various substances on cell health. The 5g bottle is a convenient size for smaller scale studies or for labs with less frequent staining requirements.