Learn More
Research Products International Corp Neutral Red Dye, 5 Grams
SDP
Catalog No. 50488850
Encompass_Preferred
Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red.
Dye Content: >90%
Catalog No. 50-488-850 Supplier Research Products International Corp Supplier No. N224005.0
Biological dye used to stain nuclei red and in testing viability of cells to incorporate and bind Neutral Red.
Dye Content: >90%