Learn More
Sigma Aldrich Fine Chemicals Biosciences Neutral Red for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0) | 553-24-2 | MFCD00012651 | 25G
Catalog No. 501805226
Encompass
Neutral Red for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0) | Mol Wt: 288.78 | 553-24-2 | MFCD00012651 | 25G
Catalog No. 50-180-5226 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7221025G
Neutral Red for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0) | Mol Wt: 288.78 | 553-24-2 | MFCD00012651 | 25G