Learn More
Medchemexpress LLC Neutral Red (IND) | 553-24-2 | 96.3% | 100 G
SDP
Catalog No. 5000410319
Encompass_Preferred
Neutral Red (IND) is an organic dye commonly used in biology and cytology laboratories. It can be used to stain living cells, secreted proteins and other molecular structures, and has a wide range of applications in cell imaging and staining. In addition, Neutral Red (IND) is widely used in industrial fields such as water treatment, food processing and paper manufacturing, for example as an indicator or colorant. Although the compound has no direct medical application, it has important application value in the fields of biology, chemistry and industry.
  • Stains living cells, secreted proteins and other molecular structures.
  • Wide range of applications in cell imaging and staining.
  • Used in industrial fields such as water treatment, food processing and paper manufacturing as an indicator or colorant.
Catalog No. 50-004-10319 Supplier Medchemexpress LLC Supplier No. HYD0166A100G
Neutral Red (IND) is an organic dye commonly used in biology and cytology laboratories. It can be used to stain living cells, secreted proteins and other molecular structures, and has a wide range of applications in cell imaging and staining. In addition, Neutral Red (IND) is widely used in industrial fields such as water treatment, food processing and paper manufacturing, for example as an indicator or colorant. Although the compound has no direct medical application, it has important application value in the fields of biology, chemistry and industry.
  • Stains living cells, secreted proteins and other molecular structures.
  • Wide range of applications in cell imaging and staining.
  • Used in industrial fields such as water treatment, food processing and paper manufacturing as an indicator or colorant.