Learn More
Crescent Chemical Co Inc NEUTRAL RED PURE
SDP
Catalog No. 50247918
Encompass_Preferred
Neutral Red pure Indicator pH 6.8 - 8.0. For use in histology and supravital staining. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 30305.01 CAS 553-24-2
Catalog No. 50-247-918 Supplier Crescent Chemical Co Inc Supplier No. SE30305.01
Neutral Red pure Indicator pH 6.8 - 8.0. For use in histology and supravital staining. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 30305.01 CAS 553-24-2