Learn More
Quality Biological Inc Neutral Red Solution
SDP
Supplier:  Quality Biological Inc 115341061
Encompass_Preferred
Neutral Red Solution is also known as Basic Red 5, C.I. 50040, toluylene red, or 3-amino-7-dimethyl amino-2-methylphenazine HCl.This formulation of neutral red contains 3.33 g/L of the cationic neutral red dye.ApplicationsHistology: Neutral red is commonly used in histological staining applications. Virology: The neutral red solution is also used in the plaque assay for the quantification of infectious particles by quantifying the plaques formed in cell culture following infection with serial dilutions of an infectious sample. Cytology: The neutral red solution application extends to the assessment of cell proliferation and cytotoxicity in the neutral red uptake assay used to quantify the number of viable cells in culture."
Catalog No. 50-288-4310