Learn More
MilliporeSigma™ Neutral Red Stain
Catalog No. M1013760025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
M1013760025 25 g
1 options
Catalog No. M1013760025 Supplier MilliporeSigma™ Supplier No. 1.01376.0025
Stains bacteriological sample material of human origin.
  • Dry dye
  • Used for preparation of staining solution
  • Applied in histology for nuclear staining, fluorescence staining, polychrome staining or for counter staining in Gram staining
  • used for supravital staining of leukocytes in blood of human origin.
TRUSTED_SUSTAINABILITY
Specifications
CAS 553-24-2
Packaging Glass Bottle
Quantity 25 g