Learn More
Scigen Neutralex™ Formalin Test Kit
Catalog No. 23730564 Shop All Scigen Inc Products
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
23-730-564 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 23-730-564 Supplier Scigen Supplier No. 4050
This item is not returnable. View return policy
20 seconds fast and reliable semiquantitative determination of residual formalin and pH in Neutralex treated formalin waste. Includes testing protocol.
Neutralex™ Aldehyde Test Kit is used to test the pH and aldehyde content of spent waste in 20 seconds. It has been designed specifically for use with Neutralex™ franchise products for the safe treatment and disposal of formalin and aldehyde waste.
Each kit is complete with a reagent solution and pH strips that are used to determine when the waste being treated has been neutralized and is safe for disposal down the drain. Users should always log their results for reporting purposes.
Neutralex™ products are certified by the California Environmental Protection Agency. The seal indicates that the formalin treatment process results in non-hazardous drain safe waste.
  • Semiquantitative determination of residual formaldehyde in treated waste, determination points 0, 10, 20 part per million (ppm)
  • Fast and reliable determination
  • 100 residual formaldehyde and 100 pH determinations
  • No sodium hydroxide reagent needed to activate formaldehyde test strip
  • Manufactured using ISO 13485 and FDA quality systems
Specifications
Description For testing Neutralex™ treated formalin waste
Type Formalin Test Kit
For Use With (Application) To determine pH and formaldehyde in Neutralex™ treated waste
Includes pH and residual formaldehyde testing procedure
Packaging Quantity 1 Ea.