Learn More
Specifications
Specifications
| Concentration | 0.1 mg/mL |
| Conjugate | SPRD (R-phycoerythrin-Cy5) |
| Formulation | Supplied in PBS/NaN3 and a stabilizing agent |
| Quantity | 0.1 mg |
| Content And Storage | Store at 2°C to 8°C; Protect from light; Do not freeze |
| For Use With (Application) | FC |
| Product Type | Avidin |
For Research Use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
