Learn More
Lifeline Cell Technology Trypsin Neutralizing Solution, 100mL
SDP
Catalog No. NC9972663
Encompass_Preferred
Trypsin is an enzyme that breaks down proteins to dissociate cells from the vessel in which they’re being cultured.
Catalog No. NC9972663 Supplier Lifeline Cell Technology Supplier No. CM0018
Trypsin is an enzyme that breaks down proteins to dissociate cells from the vessel in which they’re being cultured.