Learn More
Macherey-Nagel NEUTRALIZATION BUFFER NEU EF
Catalog No. NC2222979
Encompass_Preferred
Neutralization Buffer NEU EF to be used with NucleoBond Xtra Midi / Maxi EF kits
This item is not returnable. View return policy
Catalog No. NC2222979 Supplier Macherey-Nagel Supplier No. 740388.1000
Neutralization Buffer NEU EF to be used with NucleoBond Xtra Midi / Maxi EF kits