Learn More
Thermo Scientific™ Neutralization Solution
Catalog No. FERR1591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
240 mL
Catalog No. Quantity
FERR1591 240 mL
1 options
Catalog No. FERR1591 Supplier Thermo Scientific™ Supplier No. R1591
Add to Cart
Add to Cart
Thermo Scientific GeneJET Neutralization Solution is a component of the GeneJET Plasmid Maxiprep Kit and may be purchased separately. Provided in 240 mL aliquots.
Thermo Scientific GeneJET Neutralization Solution is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately. Provided in 240 mL aliquots.
Thermo Scientific GeneJET Neutralization Solution is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately. Provided in 240 mL aliquots.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Neutralization Solution
Product Line GeneJET
Product Type Neutralization Solution
Quantity 240 mL
For Research Use Only. Not for use in diagnostic procedures.