Learn More
Zymo Research Corporation Neutralization Buffer (Yellow), 20 ml
SDP
Catalog No. 50444226
Encompass_Preferred
Neutralization Buffer (Yellow) is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.
Catalog No. 50-444-226 Supplier Zymo Research Corporation Supplier No. D4036220
Neutralization Buffer (Yellow) is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.