Learn More
Sigma Aldrich Fine Chemicals Biosciences Neutralizing Buffer for No1L
Catalog No. 502904621
Encompass
Sigmas Neutralization Buffer for Southern procedures will neutralize the pH of the gel following depurination and denaturation treatment.
Catalog No. 50-290-4621 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N60195X1L
Sigmas Neutralization Buffer for Southern procedures will neutralize the pH of the gel following depurination and denaturation treatment.