Learn More
Kem Medical Products Corp NEUTRALIZING POWDER 24/CS
SDP
Catalog No. NC0460400
Encompass_Preferred
Kem-Safe Glutaraldehyde/OPA Neutralizing Powder 24 cont.s/cs (each cont. neutralizes one gal waste solution)
This item is not returnable. View return policy
Catalog No. NC0460400 Supplier Kem Medical Products Corp Supplier No. 9075
Kem-Safe Glutaraldehyde/OPA Neutralizing Powder 24 cont.s/cs (each cont. neutralizes one gal waste solution)