Learn More
Bioworld Neutralizing Solution, 1 L
Catalog No. 50196834
Encompass_Preferred
Neutralizing Solution Biotechnology Grade Technical Information:Contains 1.5M NaCl and 1.0M TrisConductivity (1:100)(umhos): 2300-3000pH @25C: 6.9-7.1Sterility(P/F): PassStorage: RTHazard: Irritant
Catalog No. 50-196-834 Supplier Bioworld Supplier No. 107500141
Neutralizing Solution Biotechnology Grade Technical Information:Contains 1.5M NaCl and 1.0M TrisConductivity (1:100)(umhos): 2300-3000pH @25C: 6.9-7.1Sterility(P/F): PassStorage: RTHazard: Irritant