Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Reference Material, Neutrals QC, 2 mL, Liquid

Catalog No. 50466292 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Reference Material is for chromatographic systems with a UV detector. It's a system benchmarking standard, with a neutral compound mix for confidence in results, reduced troubleshooting, and cross-laboratory comparisons. Adaptable to diverse separation methods.

  • For chromatographic systems with a UV detector
  • System benchmarking standard
  • Composed of neutral compounds
  • Adaptable to diverse separation methods
  • Separation mode: Reversed Phase
  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room Temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 24 Months
  • Shipping condition: Ambient
  • Volume: 2 mL
  • Application: Small Molecule
  • Kit includes ampule with 10 μL/mL acetone, 0.25 mg/mL naphthalene, 0.4 mg/mL acenaphthene in 50/50 acetonitrile/water, 2 mL

Catalog No. 50-466-292 Supplier Waters Corp Supplier No. 186006360
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Reference Material is for chromatographic systems with a UV detector. It's a system benchmarking standard, with a neutral compound mix for confidence in results, reduced troubleshooting, and cross-laboratory comparisons. Adaptable to diverse separation methods.

  • For chromatographic systems with a UV detector
  • System benchmarking standard
  • Composed of neutral compounds
  • Adaptable to diverse separation methods
  • Separation mode: Reversed Phase
  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room Temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 24 Months
  • Shipping condition: Ambient
  • Volume: 2 mL
  • Application: Small Molecule
  • Kit includes ampule with 10 μL/mL acetone, 0.25 mg/mL naphthalene, 0.4 mg/mL acenaphthene in 50/50 acetonitrile/water, 2 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.