Learn More
Azer Scientific Neutralyde™
SDP
Catalog No. 21402483
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
21-402-483 1 gal.
1 options
Catalog No. 21-402-483 Supplier Azer Scientific Supplier No. ES766
Add to Cart
Add to Cart
Designed to safely neutralize and dispose of formalin in the laboratory environment
  • Designed to safely neutralize and dispose of formalin in the laboratory environment without the threat of exposure to hazardous vapors
  • Treated waste may be disposed of in a sanitary drain
  • No pH adjustment is required when used properly
  • Neutralization is complete in 8 hour
  • Phosphate-free
  • One gallon of Neutralyde™ for five gallons of 10% formalin waste safe gentle reaction
Specifications
Quantity 1 gal.