Learn More
MOLECULAR DEPOT LLC Neutravidin Beads - 1 mL
Catalog No. 5000013502
Encompass_Preferred
Neutravidin Magnetic Beads are a 1perc solid suspension of 0 9-1 1 um superparamagnetic iron oxide beads covalently conjugated to Neutravidin Highly stable (no-leaching) and with ultra-low non-specific binding and high capacity these beads have been tested in several assays for the capture of biotin-labelled antigens antibodies and nucleic acids For in vitro research use only
Catalog No. 50-000-13502 Supplier MOLECULAR DEPOT LLC Supplier No. M2010003
Neutravidin Magnetic Beads are a 1perc solid suspension of 0 9-1 1 um superparamagnetic iron oxide beads covalently conjugated to Neutravidin Highly stable (no-leaching) and with ultra-low non-specific binding and high capacity these beads have been tested in several assays for the capture of biotin-labelled antigens antibodies and nucleic acids For in vitro research use only