Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NeutrAvidin Protein, FITC

Catalog No. PI31006 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31006 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31006 Supplier Invitrogen™ Supplier No. 31006
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Protein

NeutrAvidin for Western Blot, IHC, ELISA, ISH

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. The biotin-binding protein contains four identical subunits having a combined mass of 67,000-68,000 daltons. Removing the glycosylation from avidin yields NeutrAvidin Protein with a mass of 60,000 daltons. Carbohydrate-based lectin-binding is reduced to undetectable levels, yet biotin-binding affinity is retained. NeutrAvidin Protein offers the advantages of a neutral pI to minimize nonspecific adsorption, along with lysine residues that remain available for derivatization or other customized conjugation. NeutrAvidin Protein yields the lowest nonspecific binding among the known biotin binding proteins. The specific activity for biotin-binding is approximately 14 μg/mg of protein, which is near the theoretical maximum activity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot
Formulation PBS with no preservative
Name NeutrAvidin
Conjugate FITC
Cross Reactivity Tag
Purification Method Purified
Quantity 5 mg
Storage Requirements 4°C, store in dark
Regulatory Status RUO
Common Name NeutrAvidin
Product Type Protein, NeutrAvidin
Content And Storage 4°C, Upon receipt store product 4°C. Product is shipped at ambient temperature.
Form Lyophilized
Purity or Quality Grade purified
Host Species N/A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.