Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neutrophil activating peptide 2 (CXCL7) | >95.0% | 500 UG
SDP

Catalog No. 5000290476
Encompass_Preferred

Recombinant mouse NAP-2 (CXCL7) is a His-tagged CXC chemokine produced in E. coli and supplied lyophilized. It comprises residues K40-Y113 (74 amino acids) and is intended for in vitro research applications involving neutrophil chemotaxis and receptor binding.

  • Recombinant His-tagged mouse chemokine for in vitro use.
  • Contains amino acids K40-Y113 (74 aa).
  • Expressed in E. coli for high-yield production.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for storage.
  • Recommended reconstitution ≥100 μg/mL in ddH2O.
  • Stable at -20°C for long-term storage; aliquot to avoid freeze-thaw.

Catalog No. 50-002-90476 Supplier Medchemexpress LLC Supplier No. HYP75698500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse NAP-2 (CXCL7) is a His-tagged CXC chemokine produced in E. coli and supplied lyophilized. It comprises residues K40-Y113 (74 amino acids) and is intended for in vitro research applications involving neutrophil chemotaxis and receptor binding.

  • Recombinant His-tagged mouse chemokine for in vitro use.
  • Contains amino acids K40-Y113 (74 aa).
  • Expressed in E. coli for high-yield production.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for storage.
  • Recommended reconstitution ≥100 μg/mL in ddH2O.
  • Stable at -20°C for long-term storage; aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.