Learn More
ACROBiosystems Human Neutrophil defensin 1 / DEFA1 protein, Twin-Strep Tag
Catalog No. 502719130
Encompass_Preferred
Human defensins are a family of small antimicrobial proteins found predominantly in leukocytes and epithelial cells that play important roles in the innate and adaptive immune defense against microbial infection. The α-defensins, human neutrophil peptides (HNPs) 1–3, are constitutively expressed in neutrophils and are the most abundant neutrophil granule proteins.
Catalog No. 50-271-9130 Supplier ACROBiosystems Supplier No. ND1H528250UG
Human defensins are a family of small antimicrobial proteins found predominantly in leukocytes and epithelial cells that play important roles in the innate and adaptive immune defense against microbial infection. The α-defensins, human neutrophil peptides (HNPs) 1–3, are constitutively expressed in neutrophils and are the most abundant neutrophil granule proteins.