Learn More
Santa Cruz Biotechnology Neutrophil Elastase Antibody (G-2) 200 ug/ml
Catalog No. NC2955707
Encompass
Neutrophil Elastase Antibody (G-2) is a mouse monoclonal IgG1 κ Neutrophil Elastase antibody, cited in 28 publications, provided at 200 µg/mlraised against amino acids 211-267 of Neutrophil Elastase of human originNeutrophil Elastase Antibody (G-2) is recommended for detection of Neutrophil Elastase of human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC2955707 Supplier Santa Cruz Biotechnology Supplier No. SC55549
Neutrophil Elastase Antibody (G-2) is a mouse monoclonal IgG1 κ Neutrophil Elastase antibody, cited in 28 publications, provided at 200 µg/mlraised against amino acids 211-267 of Neutrophil Elastase of human originNeutrophil Elastase Antibody (G-2) is recommended for detection of Neutrophil Elastase of human origin by WB, IP, IF, IHC(P) and ELISA