Learn More
Cayman Chemical Neutrophil Elastase Inhibitor, Purity: ≥95%, CAS: 1448314-31-5, 5 mg
SDP
Catalog No. 502969952
Encompass_Preferred
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)
Catalog No. 50-296-9952 Supplier Cayman Chemical Supplier No. 149225 MG
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)