Learn More
Innovative Research Inc Human Neutrophil Elastase Purified Lyophilized 500ug (5x100ug)
Catalog No. 502036473
Encompass_Preferred
Human Neutrophil Elastase Purified Lyophilized from Innovative Research is fractionated with NaCl and treated with DNase to release DNA-bound proteins. The elastase is purified by affinity chromatography on Agarose-DAPSA-Elastin Soluble and separated from cathepsins by ion exchange chromatography on carboxymethyl-agarose. The elastase is further purified by affinity chromatography at -15°C on agarose-alanyl-alanyl-alanine. The elastase is free of cathepsin G activity when assayed with Suc-Ala-Ala-Pro-Phe-pNA.Free of cathepsin G, MPO and lysozyme..
Catalog No. 50-203-6473 Supplier Innovative Research Inc Supplier No. IHUELASD500UG
Human Neutrophil Elastase Purified Lyophilized from Innovative Research is fractionated with NaCl and treated with DNase to release DNA-bound proteins. The elastase is purified by affinity chromatography on Agarose-DAPSA-Elastin Soluble and separated from cathepsins by ion exchange chromatography on carboxymethyl-agarose. The elastase is further purified by affinity chromatography at -15°C on agarose-alanyl-alanyl-alanine. The elastase is free of cathepsin G activity when assayed with Suc-Ala-Ala-Pro-Phe-pNA.Free of cathepsin G, MPO and lysozyme..