Learn More
AdipoGen anti-Neutrophils (mouse), mAb (blocking) (Nimp-R14) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0043BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse neutrophils (surface protein mouse Ly-6G) and may recognize Ly-6Chigh cells (inflammatory monocytes). Isotype: Rat IgG2b. Clone: Nimp-R14. Applications: FACS, FUNC (Blocking), ICC, IHC. Liquid. In PBS containing 0.02% sodium azide. Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.
Catalog No. 50-205-2950