Learn More
Cayman Chemical Neutrophil Myeloperoxidase Ac
SDP
Catalog No. 502895204
Encompass_Preferred
Cayman’s Neutrophil Myeloperoxidase Activity Assay kit utilizes 3,3’,5,5’-tetramethyl-benzidine (TMB) as a chromogenic substrate, which upon reacting with MPO, yields a blue color detectable by its absorbance at 650 nm. The color intensity is proportional to the amount of MPO in the sample. Reagents needed to isolate neutrophils from whole blood are included in the kit, as is PMA, which is known to stimulate MPO release from neutrophils. A specific inhibitor for MPO, 4-aminobenzhydrazide (ABH), is also included in the kit for verifying the specificity of the assay.
Catalog No. 50-289-5204 Supplier Cayman Chemical Supplier No. 6006201 EA
Cayman’s Neutrophil Myeloperoxidase Activity Assay kit utilizes 3,3’,5,5’-tetramethyl-benzidine (TMB) as a chromogenic substrate, which upon reacting with MPO, yields a blue color detectable by its absorbance at 650 nm. The color intensity is proportional to the amount of MPO in the sample. Reagents needed to isolate neutrophils from whole blood are included in the kit, as is PMA, which is known to stimulate MPO release from neutrophils. A specific inhibitor for MPO, 4-aminobenzhydrazide (ABH), is also included in the kit for verifying the specificity of the assay.