Learn More
AdipoGen Neutrophilsms mAb Nimp-R14
SDP
Supplier:  AdipoGen AG20B0043PFC500
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse neutrophils surface protein mouse Ly-6G and may recognize Ly-6Chigh cells inflammatory monocytes. Isotype Rat IgG2b. Clone Nimp-R14. Applications FACS, FUNC Blocking, ICC, IHC. Liquid. In PBS containing 10% glycerol. Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.
Catalog No. 50-256-3378