Learn More
Medchemexpress LLC Nevadensin | 10176-66-6 | 344.32 | 5 MG
SDP
Catalog No. 5000416558
Encompass_Preferred
Nevadensin (Lysionotin) is a natural flavonoid that acts as a selective human carboxylesterase 1 (hCE1) inhibitor with an IC50 of 2.64 μM, showing more selectivity for hCE1 than hCE2 (IC50 of 132.8 μM). It can induce apoptosis and DNA damage in cancer cells. This compound exhibits a range of pharmacological effects, including anti-inflammatory, anti-tumour, anti-hypertensive, anti-tubercular, antitussive, antioxidant, and anti-microbial activities.
  • Natural flavonoid
  • Selective human carboxylesterase 1 (hCE1) inhibitor
  • Induces apoptosis and DNA damage in cancer cells
  • Possesses anti-inflammatory activity
  • Exhibits anti-tumour effects
  • Has anti-hypertensive properties
  • Demonstrates anti-tubercular activity
  • Acts as an antitussive
  • Functions as an antioxidant
  • Shows anti-microbial activities
Catalog No. 50-004-16558 Supplier Medchemexpress LLC Supplier No. HYN13775MG
Nevadensin (Lysionotin) is a natural flavonoid that acts as a selective human carboxylesterase 1 (hCE1) inhibitor with an IC50 of 2.64 μM, showing more selectivity for hCE1 than hCE2 (IC50 of 132.8 μM). It can induce apoptosis and DNA damage in cancer cells. This compound exhibits a range of pharmacological effects, including anti-inflammatory, anti-tumour, anti-hypertensive, anti-tubercular, antitussive, antioxidant, and anti-microbial activities.
  • Natural flavonoid
  • Selective human carboxylesterase 1 (hCE1) inhibitor
  • Induces apoptosis and DNA damage in cancer cells
  • Possesses anti-inflammatory activity
  • Exhibits anti-tumour effects
  • Has anti-hypertensive properties
  • Demonstrates anti-tubercular activity
  • Acts as an antitussive
  • Functions as an antioxidant
  • Shows anti-microbial activities