Learn More
Medchemexpress LLC Nevadensin | 10176-66-6 | MFCD08689947 | 344.32 | 20 MG
SDP
Catalog No. 5000415747
Encompass_Preferred
Nevadensin is a natural flavonoid and a selective human carboxylesterase 1 (hCE1) inhibitor. It demonstrates greater selectivity for hCE1 over hCE2.
  • Induces apoptosis and DNA damage in cancer cells.
  • Possesses anti-inflammatory, anti-tumor, anti-hypertensive, anti-tubercular, antitussive, antioxidant, and anti-microbial activities.
  • Inhibits hCE1 with an IC50 of 2.64 μM.
Catalog No. 50-004-15747 Supplier Medchemexpress LLC Supplier No. HYN137720MG
Nevadensin is a natural flavonoid and a selective human carboxylesterase 1 (hCE1) inhibitor. It demonstrates greater selectivity for hCE1 over hCE2.
  • Induces apoptosis and DNA damage in cancer cells.
  • Possesses anti-inflammatory, anti-tumor, anti-hypertensive, anti-tubercular, antitussive, antioxidant, and anti-microbial activities.
  • Inhibits hCE1 with an IC50 of 2.64 μM.