Learn More
Medchemexpress LLC Nevadensin (5,7-dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)chromen-4-one) | 10176-66-6 | MFCD08689947 | 99.9% | 344.3 g/mol | C18H16O7 | 10 MG
SDP
Catalog No. 5000295723
Encompass_Preferred
Nevadensin (5,7-dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)chromen-4-one) is the analytical standard of a naturally occurring flavone used in biochemical and pharmacological research. It is commonly used as a reference material for assay calibration, method development, and inhibitor screening.
  • Analytical reference standard suitable for assay calibration.
  • Reported human carboxylesterase 1 (hCE1) inhibitory activity for research use.
  • High chemical purity for reliable analytical results.
  • Stable solid form with defined molecular identifiers for traceability.
  • Compact, small-quantity format for laboratory testing and method validation.
Catalog No. 50-002-95723 Supplier Medchemexpress LLC Supplier No. HYN137710MG
Nevadensin (5,7-dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)chromen-4-one) is the analytical standard of a naturally occurring flavone used in biochemical and pharmacological research. It is commonly used as a reference material for assay calibration, method development, and inhibitor screening.
  • Analytical reference standard suitable for assay calibration.
  • Reported human carboxylesterase 1 (hCE1) inhibitory activity for research use.
  • High chemical purity for reliable analytical results.
  • Stable solid form with defined molecular identifiers for traceability.
  • Compact, small-quantity format for laboratory testing and method validation.