Learn More
Medchemexpress LLC Nevanimibe hydrochloride | 133825-81-7 | 99.5% | 458.08 | C27H40ClN3O | 10 MG
SDP
Catalog No. 5000215489
Encompass_Preferred
Nevanimibe hydrochloride is the hydrochloride salt of Nevanimibe, an orally active, selective inhibitor of acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) with demonstrated activity in preclinical studies. It is provided for research use in solid and solution forms and has been characterized for potency and basic physicochemical properties.
  • Selective ACAT1 inhibitor with low-nanomolar potency
  • Demonstrated activity against ACAT2 in cellular assays
  • Induces apoptosis in cell models relevant to adrenocortical tissues
  • Chemical formula C27H40ClN3O and molecular weight 458.08
  • Available as solid and DMSO solution formats for research use
Catalog No. 50-002-15489 Supplier Medchemexpress LLC Supplier No. HY100399A10MG
Nevanimibe hydrochloride is the hydrochloride salt of Nevanimibe, an orally active, selective inhibitor of acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) with demonstrated activity in preclinical studies. It is provided for research use in solid and solution forms and has been characterized for potency and basic physicochemical properties.
  • Selective ACAT1 inhibitor with low-nanomolar potency
  • Demonstrated activity against ACAT2 in cellular assays
  • Induces apoptosis in cell models relevant to adrenocortical tissues
  • Chemical formula C27H40ClN3O and molecular weight 458.08
  • Available as solid and DMSO solution formats for research use