Learn More
Medchemexpress LLC Nevanimibe hydrochloride | 133825-81-7 | 99.5% | 458.08 g·mol⁻¹ | C27H40ClN3O | 100 MG
SDP
Catalog No. 5000214054
Encompass_Preferred
Nevanimibe hydrochloride is an orally active, selective inhibitor of acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) used in preclinical research. It has low-nanomolar activity against ACAT1 and has been investigated in cell models for effects on cholesterol metabolism and adrenocortical cancer.
  • Selective ACAT1 inhibitor with EC50 ≈ 9 nM.
  • Orally active small molecule suitable for in vitro and in vivo research.
  • High chemical purity (~99.5%) for reliable experimental results.
  • Available as solid and as 10 mM solution in DMSO for flexible dosing.
  • Characterized by CAS 133825-81-7 and molecular weight 458.08 g·mol⁻¹.
Catalog No. 50-002-14054 Supplier Medchemexpress LLC Supplier No. HY100399A100MG
Nevanimibe hydrochloride is an orally active, selective inhibitor of acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) used in preclinical research. It has low-nanomolar activity against ACAT1 and has been investigated in cell models for effects on cholesterol metabolism and adrenocortical cancer.
  • Selective ACAT1 inhibitor with EC50 ≈ 9 nM.
  • Orally active small molecule suitable for in vitro and in vivo research.
  • High chemical purity (~99.5%) for reliable experimental results.
  • Available as solid and as 10 mM solution in DMSO for flexible dosing.
  • Characterized by CAS 133825-81-7 and molecular weight 458.08 g·mol⁻¹.