Learn More
Medchemexpress LLC Nevanimibe hydrochloride | 133825-81-7 | MFCD00899568 | 99.5% | 458.08 g/mol | C27H40ClN3O | 1 ML
SDP
Catalog No. 5000215477
Encompass_Preferred
Nevanimibe hydrochloride is the hydrochloride salt of nevanimibe, a selective acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for in vitro research use.
  • Selective ACAT1 inhibitor with low-nanomolar activity
  • Supplied as a ready-to-use 10 mM solution in DMSO
  • High purity (~99.5%) suitable for biochemical and cell-based assays
  • Molecular weight 458.08 g/mol aids solution preparation calculations
  • Available in solid and solution formats for flexibility in workflows
Catalog No. 50-002-15477 Supplier Medchemexpress LLC Supplier No. HY100399A10MM 1ML
Nevanimibe hydrochloride is the hydrochloride salt of nevanimibe, a selective acyl-coenzyme A:cholesterol O-acyltransferase 1 (ACAT1) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for in vitro research use.
  • Selective ACAT1 inhibitor with low-nanomolar activity
  • Supplied as a ready-to-use 10 mM solution in DMSO
  • High purity (~99.5%) suitable for biochemical and cell-based assays
  • Molecular weight 458.08 g/mol aids solution preparation calculations
  • Available in solid and solution formats for flexibility in workflows