Learn More
Medchemexpress LLC HY-10570 10mg Medchemexpress, Nevirapine CAS:129618-40-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1057010MG
Encompass_Preferred
Medchemexpress, HY-10570 10mg Nevirapine CAS:129618-40-2 Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used to treat and prevent HIV/AIDS; with a Ki of 270 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8290