Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nevirapine | 129618-40-2 | MFCD00866928 | 99.9% | 266.30 | C15H14N4O | 200 MG
SDP

Catalog No. 5000240219
Encompass_Preferred

Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used as a research reagent to study reverse transcriptase inhibition and antiviral activity. It is characterized for analytical and biological assays, with documentation for solubility and handling in product literature.

  • Non-nucleoside reverse transcriptase inhibitor activity (reported Ki ~270 μM).
  • Suitable for in vitro HIV research and biochemical assays.
  • High purity for research use (COA reports 99.93% by LCMS).
  • Chemical formula C15H14N4O; molecular weight 266.30.
  • Solubility and storage information provided in product documentation for proper handling.

Catalog No. 50-002-40219 Supplier Medchemexpress LLC Supplier No. HY10570200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nevirapine is a non-nucleoside inhibitor of HIV-1 reverse transcriptase used as a research reagent to study reverse transcriptase inhibition and antiviral activity. It is characterized for analytical and biological assays, with documentation for solubility and handling in product literature.

  • Non-nucleoside reverse transcriptase inhibitor activity (reported Ki ~270 μM).
  • Suitable for in vitro HIV research and biochemical assays.
  • High purity for research use (COA reports 99.93% by LCMS).
  • Chemical formula C15H14N4O; molecular weight 266.30.
  • Solubility and storage information provided in product documentation for proper handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.