Learn More
Medchemexpress LLC Nevirapine-d4 | 1051418-95-1 | ≥99.0% | 270.32 | C15H10D4N4O | 1 MG
SDP
Catalog No. 5000240517
Encompass_Preferred
Nevirapine-d4 is a deuterium-labeled analogue of nevirapine supplied as a research-grade analytical standard. It is intended for use as an internal standard in LC-MS/MS assays, pharmacokinetic and metabolic studies, and analytical method development where isotopically labeled reference material is required. Supplied as a 1 mg solid with high deuterium incorporation.
Catalog No. 50-002-40517 Supplier Medchemexpress LLC Supplier No. HY10570S1MG
Add to Cart
Add to Cart
Nevirapine-d4 is a deuterium-labeled analogue of nevirapine supplied as a research-grade analytical standard. It is intended for use as an internal standard in LC-MS/MS assays, pharmacokinetic and metabolic studies, and analytical method development where isotopically labeled reference material is required. Supplied as a 1 mg solid with high deuterium incorporation.