Learn More
Nevirapine Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 287980-85-2 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C15H16N4 O |
| Synonym | 5,11-Dihydro-4-methyl-11-propyl-6 H-dipyrido[3,2-b:2′,3′-e][1,4 ]diazepin-6-one; 5,11-DIHYDRO-6 H-11-PROPYL-4 -METHYL-DIPYRIDO [3,2-b:2′,3′-e][1,4 ] DIAZEPIN-6-ONE |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 268.31 |
| Grade | Certified Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.