Learn More
Selleck Chemical LLC Nevirapine 5mg 129618-40-2 NSC 641530
SDP
Supplier:  Selleck Chemical LLC S17425MG
Encompass_Preferred
Nevirapine (NSC 641530) is a non-nucleoside reverse transcriptase inhibitor (NNRTI) used to treat HIV-1 infection and AIDS. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-612-6